diexo-3-tert-Butoxycarbonylamino-7-oxa-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid
Lieferant: Apollo Scientific
OR309425-250MG
OR309425-500MG
APOSOR309425-250MGEA
185
EUR
APOSOR309425-250MG
APOSOR309425-500MG
diexo-3-tert-Butoxycarbonylamino-7-oxa-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid
diexo-3-tert-Butoxycarbonylamino-7-oxa-bicyclo[2.2.1]hept-5-ene-2-carboxylic acid
Formel:
C₁₂H₁₇NO₅ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07783749 CAS-Nummer: 148257-14-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...