1-Amino-2-phenyl-propan-2-ol

Lieferant: Apollo Scientific

OR309430-1G OR309430-5G
APOSOR309430-1GEA 155 EUR
APOSOR309430-1G APOSOR309430-5G
1-Amino-2-phenyl-propan-2-ol
1-Amino-2-phenyl-propan-2-ol
Formel: C₉H₁₃NO
Molecular Weight: 151.206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114539
CAS-Nummer: 1017418-99-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR