5-Amino-1-methyl-2,3-dihydro-1H-1,3-benzodiazol-2-one
Lieferant: Apollo Scientific
|
Achtung
|
OR310077-1G
OR310077-500MG
APOSOR310077-1GEA
480
EUR
APOSOR310077-1G
APOSOR310077-500MG
5-Amino-1-methyl-2,3-dihydro-1H-1,3-benzodiazol-2-one
5-Amino-1-methyl-2,3-dihydro-1H-1,3-benzodiazol-2-one
Formel:
C₈H₉N₃O Molecular Weight: 163.177 g/mol Schmelzpunkt: 222…224 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD20691260 CAS-Nummer: 54732-89-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...