2-Hydroxy-5-isopropylbenzonitrile

Lieferant: Apollo Scientific
Achtung

OR31008-1G OR31008-250MG
APOSOR31008-1GEA 812 EUR
APOSOR31008-1G APOSOR31008-250MG
2-Hydroxy-5-isopropylbenzonitrile
2-Hydroxy-5-isopropylbenzonitrile
Formel: C₁₀H₁₁NO
Molecular Weight: 161,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16876983
CAS-Nummer: 143912-49-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR