N-[3-(N',N'-Dimethylamino)propyl]benzamide-4-boronic acid, pinacol ester

Lieferant: Apollo Scientific

Synonyme: 4-[[3-(Dimethylamino)propyl]carbamoyl]phenylboronic Acid Pinacol Ester

OR310098-1G OR310098-500MG
APOSOR310098-1GEA 270 EUR
APOSOR310098-1G APOSOR310098-500MG
N-[3-(N',N'-Dimethylamino)propyl]benzamide-4-boronic acid, pinacol ester
N-[3-(N',N'-Dimethylamino)propyl]benzamide-4-boronic acid, pinacol ester
Formel: C₁₈H₂₉BN₂O₃
Molecular Weight: 332.245 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05864306
CAS-Nummer: 832114-10-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR