3,4-Dichlorofuran-2(5H)-one

Lieferant: Apollo Scientific
Achtung

OR31019-1G OR31019-5G
APOSOR31019-1GEA 79.9 EUR
APOSOR31019-1G APOSOR31019-5G
3,4-Dichlorofuran-2(5H)-one
3,4-Dichlorofuran-2(5H)-one
Formel: C₄H₂Cl₂O₂
Molecular Weight: 152,963 g/mol
Schmelzpunkt: 48…52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02751908
CAS-Nummer: 62674-12-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR