4-[(5-Bromo-1,3,4-thiadiazol-2-yl)oxy]phenol
Lieferant: Apollo Scientific
OR310190-1G
OR310190-500MG
APOSOR310190-1GEA
894
EUR
APOSOR310190-1G
APOSOR310190-500MG
4-[(5-Bromo-1,3,4-thiadiazol-2-yl)oxy]phenol
4-[(5-Bromo-1,3,4-thiadiazol-2-yl)oxy]phenol
Formel:
C₈H₅BrN₂O₂S Molecular Weight: 273,107 g/mol Schmelzpunkt: 167…169 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD21605750 CAS-Nummer: 946884-41-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...