4-Methyl-6-nitro-2H-1,3-benzodioxole-5-carboxylic acid

Lieferant: Apollo Scientific

OR310327-1G OR310327-500MG
APOSOR310327-1GEA 671 EUR
APOSOR310327-1G APOSOR310327-500MG
4-Methyl-6-nitro-2H-1,3-benzodioxole-5-carboxylic acid
4-Methyl-6-nitro-2H-1,3-benzodioxole-5-carboxylic acid
Formel: C₉H₇NO₆
Molecular Weight: 225.155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26523308
CAS-Nummer: 861588-35-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR