Methyl (3S,4S)-1-benzyl-4-methylpyrrolidine-3-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: (3S,4S)-Methyl 1-benzyl-4-methylpyrrolidine-3-carboxylate, (3S,4S)-1-Benzyl-3-(methoxycarbonyl)-4-methylpyrrolidine

OR310342-1G OR310342-500MG
APOSOR310342-1GEA 671 EUR
APOSOR310342-1G APOSOR310342-500MG
Methyl (3S,4S)-1-benzyl-4-methylpyrrolidine-3-carboxylate
Methyl (3S,4S)-1-benzyl-4-methylpyrrolidine-3-carboxylate
Formel: C₁₄H₁₉NO₂
Molecular Weight: 233,306 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22420558
CAS-Nummer: 181114-98-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR