2-Oxo-2-(3,4,5-trimethoxyphenyl)acetic acid

Lieferant: Apollo Scientific

OR310349-1G OR310349-500MG
APOSOR310349-1GEA 671 EUR
APOSOR310349-1G APOSOR310349-500MG
2-Oxo-2-(3,4,5-trimethoxyphenyl)acetic acid
2-Oxo-2-(3,4,5-trimethoxyphenyl)acetic acid
Formel: C₁₁H₁₂O₆
Molecular Weight: 240,209 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03844682
CAS-Nummer: 88755-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR