2-(3-Bromo-5-chlorophenyl)acetonitrile 95%

Lieferant: Apollo Scientific

OR31036-1G OR31036-250MG OR31036-5G
APOSOR31036-1GEA 120 EUR
APOSOR31036-1G APOSOR31036-250MG APOSOR31036-5G
2-(3-Bromo-5-chlorophenyl)acetonitrile 95%
2-(3-Bromo-5-chlorophenyl)acetonitrile
Formel: C₈H₅BrClN
Molecular Weight: 230,489 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13195464
CAS-Nummer: 1056454-88-6
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR