4-Chloro-1H-pyrazol-3-amine

Lieferant: Apollo Scientific
Achtung

OR310366-1G
APOSOR310366-1GEA 115 EUR
APOSOR310366-1G
4-Chloro-1H-pyrazol-3-amine
4-Chloro-1H-pyrazol-3-amine
Formel: C₃H₄ClN₃
Molecular Weight: 117,537 g/mol
Schmelzpunkt: 119…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03419764
CAS-Nummer: 54301-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR