4-(1-Methylethyl)phenylethyne

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Ethynyl-4-isopropylbenzene

OR31041-10G OR31041-1G OR31041-5G
APOSOR31041-10GEA 190 EUR
APOSOR31041-10G APOSOR31041-1G APOSOR31041-5G
4-(1-Methylethyl)phenylethyne
4-(1-Methylethyl)phenylethyne
Formel: C₁₁H₁₂
Molecular Weight: 144,213 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03839988
CAS-Nummer: 23152-99-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR