(3-Iodo-2-methylphenyl)methanol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-iodo-2-methylbenzyl alcohol

OR310415-1G OR310415-5G OR310415-10G
APOSOR310415-1GEA 91.5 EUR
APOSOR310415-1G APOSOR310415-5G APOSOR310415-10G
(3-Iodo-2-methylphenyl)methanol 95%
(3-Iodo-2-methylphenyl)methanol
Formel: C₈H₉IO
Molecular Weight: 248,061 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18397813
CAS-Nummer: 76350-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR