Methyl 6-amino-1-benzothiophene-2-carboxylate

Lieferant: Apollo Scientific

Synonyme: Methyl 6-aminobenzo[b]thiophene-2-carboxylate

OR310440-10MG OR310440-50MG
APOSOR310440-10MGEA 197 EUR
APOSOR310440-10MG APOSOR310440-50MG
Methyl 6-amino-1-benzothiophene-2-carboxylate
Methyl 6-amino-1-benzothiophene-2-carboxylate
Formel: C₁₀H₉NO₂S
Molecular Weight: 207.249 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11840222
CAS-Nummer: 57907-49-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR