N-Methyl-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
OR310476-1G
APOSOR310476-1GEA
92.8
EUR
APOSOR310476-1G
N-Methyl-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
N-Methyl-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Formel:
C₁₄H₂₀BNO₃ Molecular Weight: 261,124 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17015821 CAS-Nummer: 1197171-76-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...