Phenyl N-(propan-2-yl)carbamate

Lieferant: Apollo Scientific

OR310625-5G OR310625-1G
APOSOR310625-5GEA 806 EUR
APOSOR310625-5G APOSOR310625-1G
Phenyl N-(propan-2-yl)carbamate
Phenyl N-(propan-2-yl)carbamate
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179,216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00276276
CAS-Nummer: 17614-10-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR