trans-2,5-Bis-(hydroxymethyl)-1,4-dioxane

Lieferant: Apollo Scientific
Achtung

OR310891-250MG OR310891-100MG
APOSOR310891-250MGEA 606 EUR
APOSOR310891-250MG APOSOR310891-100MG
trans-2,5-Bis-(hydroxymethyl)-1,4-dioxane
trans-2,5-Bis-(hydroxymethyl)-1,4-dioxane
Formel: C₆H₁₂O₄
Molecular Weight: 148.157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16659995
CAS-Nummer: 87133-52-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR