4-Chloro-3-methoxybenzyl bromide

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-(Bromomethyl)-1-chloro-2-methoxybenzene

OR310902-250MG
APOSOR310902-250MGEA 97.3 EUR
APOSOR310902-250MG
4-Chloro-3-methoxybenzyl bromide
4-Chloro-3-methoxybenzyl bromide
Formel: C₈H₈BrClO
Molecular Weight: 235,506 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09907927
CAS-Nummer: 103347-14-4
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR