tert-Butyl 6-methoxy-5-nitro-1,2,3,4-tetrahydroisoquinoline-2-carboxylate

Lieferant: Apollo Scientific

OR310927-1G OR310927-500MG
APOSOR310927-1GEA 1070 EUR
APOSOR310927-1G APOSOR310927-500MG
tert-Butyl 6-methoxy-5-nitro-1,2,3,4-tetrahydroisoquinoline-2-carboxylate
tert-Butyl 6-methoxy-5-nitro-1,2,3,4-tetrahydroisoquinoline-2-carboxylate
Formel: C₁₅H₂₀N₂O₅
Molecular Weight: 308.33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28101535
CAS-Nummer: 921224-63-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR