3-(Benzenesulphonyl)-8-nitroquinoline

Lieferant: Apollo Scientific

OR310931-1G OR310931-500MG
APOSOR310931-1GEA 894 EUR
APOSOR310931-1G APOSOR310931-500MG
3-(Benzenesulphonyl)-8-nitroquinoline
3-(Benzenesulphonyl)-8-nitroquinoline
Formel: C₁₅H₁₀N₂O₄S
Molecular Weight: 314,316 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09261163
CAS-Nummer: 607743-07-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR