Methyl 6-bromo-3-hydroxy-2-methylbenzoate

Lieferant: Apollo Scientific
Achtung

OR310936-1G OR310936-500MG
APOSOR310936-1GEA 671 EUR
APOSOR310936-1G APOSOR310936-500MG
Methyl 6-bromo-3-hydroxy-2-methylbenzoate
Methyl 6-bromo-3-hydroxy-2-methylbenzoate
Formel: C₉H₉BrO₃
Molecular Weight: 245,07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16660005
CAS-Nummer: 55289-15-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR