N-[Bis(methylsulphanyl)methylidene]-4-chlorobenzene-1-sulphonamide

Lieferant: Apollo Scientific

OR310954-1G OR310954-500MG
APOSOR310954-1GEA 538 EUR
APOSOR310954-1G APOSOR310954-500MG
N-[Bis(methylsulphanyl)methylidene]-4-chlorobenzene-1-sulphonamide
N-[Bis(methylsulphanyl)methylidene]-4-chlorobenzene-1-sulphonamide
Formel: C₉H₁₀ClNO₂S₃
Molecular Weight: 295.829 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28101539
CAS-Nummer: 13068-12-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR