1-Bromo-2-(2-bromoethyl)benzene 95%

Lieferant: Apollo Scientific
Achtung

OR310964-1G OR310964-500MG OR310964-250MG
APOSOR310964-1GEA 132 EUR
APOSOR310964-1G APOSOR310964-500MG APOSOR310964-250MG
1-Bromo-2-(2-bromoethyl)benzene 95%
1-Bromo-2-(2-bromoethyl)benzene
Formel: C₈H₈Br₂
Molecular Weight: 263,957 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00027929
CAS-Nummer: 1074-15-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR