3-Benzoyl-6,7-dimethoxy-1,4-dihydroquinolin-4-one

Lieferant: Apollo Scientific

OR310995-1G
APOSOR310995-1GEA 780 EUR
APOSOR310995-1G
3-Benzoyl-6,7-dimethoxy-1,4-dihydroquinolin-4-one
3-Benzoyl-6,7-dimethoxy-1,4-dihydroquinolin-4-one
Formel: C₁₈H₁₅NO₄
Molecular Weight: 309,316 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19104962
CAS-Nummer: 1325303-43-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR