N-(2-Hydroxypropyl)-2-methylprop-2-enamide
Lieferant: Apollo Scientific
Synonyme:
N-(2-Hydroxypropyl)methacrylamide
OR311087-5G
APOSOR311087-5GEA
82.8
EUR
APOSOR311087-5G
N-(2-Hydroxypropyl)-2-methylprop-2-enamide
N-(2-Hydroxypropyl)-2-methylprop-2-enamide
Formel:
C₇H₁₃NO₂ Molecular Weight: 143,184 g/mol Schmelzpunkt: 70 °C Dichte: 1,002 g/cm³ (25 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00080531 CAS-Nummer: 21442-01-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...