9H-Fluoren-9-ylmethyl N-[(1S)-1-[methoxy(methyl)carbamoyl]-3-[(triphenylmethyl)carbamoyl]propyl]carbamate
Lieferant: Apollo Scientific
OR311113-1G
OR311113-500MG
APOSOR311113-1GEA
336
EUR
APOSOR311113-1G
APOSOR311113-500MG
9H-Fluoren-9-ylmethyl N-[(1S)-1-[methoxy(methyl)carbamoyl]-3-[(triphenylmethyl)carbamoyl]propyl]carbamate
9H-Fluoren-9-ylmethyl N-[(1S)-1-[methoxy(methyl)carbamoyl]-3-[(triphenylmethyl)carbamoyl]propyl]carbamate
Formel:
C₄₁H₃₉N₃O₅ Molecular Weight: 653,765 g/mol Schmelzpunkt: 79 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD28139665 CAS-Nummer: 474123-46-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...