2-(4-bromophenyl)cyclopropane-1-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(4-Bromophenyl)cyclopropanecarboxylic acid
OR311192-1G
OR311192-250MG
APOSOR311192-1GEA
416
EUR
APOSOR311192-1G
APOSOR311192-250MG
2-(4-bromophenyl)cyclopropane-1-carboxylic acid
2-(4-bromophenyl)cyclopropane-1-carboxylic acid
Formel:
C₁₀H₉BrO₂ Molecular Weight: 241,081 g/mol Schmelzpunkt: 122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03196280 CAS-Nummer: 77255-26-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...