[(Dimethylaminomethylidene)amino]-3H-1,2,4-dithiazoline-3-thione
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N,N-Dimethyl-N'-(3-thioxo-3H-1,2,4-dithiazol-5-yl)formimidamide
OR311272-1G
OR311272-5G
APOSOR311272-1GEA
21.9
EUR
APOSOR311272-1G
APOSOR311272-5G
[(Dimethylaminomethylidene)amino]-3H-1,2,4-dithiazoline-3-thione
[(Dimethylaminomethylidene)amino]-3H-1,2,4-dithiazoline-3-thione
Formel:
C₅H₇N₃S₃ Molecular Weight: 205,324 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19704304 CAS-Nummer: 1192027-04-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...