3-(Hydroxymethyl)bicyclo[1.1.1]pentane-1-carbonitrile

Lieferant: Apollo Scientific
Achtung

OR312131-1G OR312131-250MG
APOSOR312131-1GEA 1640 EUR
APOSOR312131-1G APOSOR312131-250MG
3-(Hydroxymethyl)bicyclo[1.1.1]pentane-1-carbonitrile
3-(Hydroxymethyl)bicyclo[1.1.1]pentane-1-carbonitrile
Formel: C₇H₉NO
Molecular Weight: 123.152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27664953
CAS-Nummer: 1370705-39-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR