3-Phenylbicyclo[1.1.1]pentane-1-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Carboxy-3-phenylbicyclo[1.1.1]pentane
OR312133-250MG
OR312133-500MG
OR312133-100MG
APOSOR312133-250MGEA
354
EUR
APOSOR312133-250MG
APOSOR312133-500MG
APOSOR312133-100MG
3-Phenylbicyclo[1.1.1]pentane-1-carboxylic acid
3-Phenylbicyclo[1.1.1]pentane-1-carboxylic acid
Formel:
C₁₂H₁₂O₂ Molecular Weight: 188,222 g/mol Schmelzpunkt: 176…177 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00101889 CAS-Nummer: 83249-04-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...