6-Azaspiro[2.5]octane hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-azaspiro[2.5]octane hydrochloride
OR312258-5G
APOSOR312258-5GEA
111
EUR
APOSOR312258-5G
6-Azaspiro[2.5]octane hydrochloride
6-Azaspiro[2.5]octane hydrochloride
Formel:
C₇H₁₃N·ClH Molecular Weight: 147,646 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14581272 CAS-Nummer: 1037834-62-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...