3-(Bicyclo[1.1.1]pentan-1-yl)aniline 95%
Lieferant: Apollo Scientific
OR312393-1G
OR312393-250MG
APOSOR312393-1GEA
2410
EUR
APOSOR312393-1G
APOSOR312393-250MG
3-(Bicyclo[1.1.1]pentan-1-yl)aniline 95%
3-(Bicyclo[1.1.1]pentan-1-yl)aniline
Formel:
C₁₁H₁₃N Molecular Weight: 159,228 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27996107 CAS-Nummer: 1823935-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...