4-(Bicyclo[1.1.1]pentan-1-yl)benzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR312395-250MG
OR312395-100MG
APOSOR312395-250MGEA
782
EUR
APOSOR312395-250MG
APOSOR312395-100MG
4-(Bicyclo[1.1.1]pentan-1-yl)benzoic acid
4-(Bicyclo[1.1.1]pentan-1-yl)benzoic acid
Formel:
C₁₂H₁₂O₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD27996109 CAS-Nummer: 1823331-06-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...