4-(Bicyclo[1.1.1]pentan-1-yl)phenol

Lieferant: Apollo Scientific
Achtung

OR312398-250MG OR312398-1G
APOSOR312398-250MGEA 499 EUR
APOSOR312398-250MG APOSOR312398-1G
4-(Bicyclo[1.1.1]pentan-1-yl)phenol
4-(Bicyclo[1.1.1]pentan-1-yl)phenol
Formel: C₁₁H₁₂O
Molecular Weight: 160,212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27996112
CAS-Nummer: 1823939-03-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR