3-(4-Bromophenyl)bicyclo[1.1.1]pentane-1-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

OR312431-250MG OR312431-100MG
APOSOR312431-250MGEA 0 EUR
APOSOR312431-250MG APOSOR312431-100MG
3-(4-Bromophenyl)bicyclo[1.1.1]pentane-1-carboxylic acid 95%
3-(4-Bromophenyl)bicyclo[1.1.1]pentane-1-carboxylic acid
Formel: C₁₂H₁₁BrO₂
Molecular Weight: 267,119 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD29037322
CAS-Nummer: 1980054-39-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR