3-(4-Bromophenyl)bicyclo[1.1.1]pentan-1-amine 95%

Lieferant: Apollo Scientific

OR312432-250MG OR312432-100MG
APOSOR312432-250MGEA 1540 EUR
APOSOR312432-250MG APOSOR312432-100MG
3-(4-Bromophenyl)bicyclo[1.1.1]pentan-1-amine 95%
3-(4-Bromophenyl)bicyclo[1.1.1]pentan-1-amine
Formel: C₁₁H₁₂BrN
Molecular Weight: 238.124 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29037323
CAS-Nummer: 1935352-56-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR