3-Cyanobicyclo[1.1.1]pentane-1-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-cyanobicyclo[1.1.1]pentane-1-carboxylic acid

OR312449-1G OR312449-250MG
APOSOR312449-1GEA 2170 EUR
APOSOR312449-1G APOSOR312449-250MG
3-Cyanobicyclo[1.1.1]pentane-1-carboxylic acid 97%
3-Cyanobicyclo[1.1.1]pentane-1-carboxylic acid
Formel: C₇H₇NO₂
Molecular Weight: 137.136 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD19229172
CAS-Nummer: 83249-02-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR