4-((tert-Butoxycarbonyl)amino)bicyclo[2.2.2]octane-1-carboxylic acid

Lieferant: Apollo Scientific

OR312487-1G OR312487-250MG
APOSOR312487-1GEA 163 EUR
APOSOR312487-1G APOSOR312487-250MG
4-((tert-Butoxycarbonyl)amino)bicyclo[2.2.2]octane-1-carboxylic acid
4-((tert-Butoxycarbonyl)amino)bicyclo[2.2.2]octane-1-carboxylic acid
Formel: C₁₄H₂₃NO₄
Molecular Weight: 269,337 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22577789
CAS-Nummer: 863304-76-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR