Dimethyl bicyclo[2.2.1]heptane-1,4-dicarboxylate
Lieferant: Apollo Scientific
OR312506-1G
OR312506-250MG
APOSOR312506-1GEA
130
EUR
APOSOR312506-1G
APOSOR312506-250MG
Dimethyl bicyclo[2.2.1]heptane-1,4-dicarboxylate
Dimethyl bicyclo[2.2.1]heptane-1,4-dicarboxylate
Formel:
C₁₁H₁₆O₄ Molecular Weight: 212,242 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23701447 CAS-Nummer: 15448-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...