Dimethyl bicyclo[2.1.1]hexane-1,4-dicarboxylate
Lieferant: Apollo Scientific
OR312518-1G OR312518-250MG
APOSOR312518-1GEA 722 EUR
APOSOR312518-1G APOSOR312518-250MG
Dimethyl bicyclo[2.1.1]hexane-1,4-dicarboxylate
Dimethyl bicyclo[2.1.1]hexane-1,4-dicarboxylate
Formel: C₁₀H₁₄O₄
Molecular Weight: 198.216 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22371985
CAS-Nummer: 42145-38-0