8-Benzyl-1,3,8-triaza-spiro[4.5]decan-4-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 8-Benzyl-1,3,8-triazaspiro[4.5]decan-4-one

OR313002-1G OR313002-250MG
APOSOR313002-1GEA 235 EUR
APOSOR313002-1G APOSOR313002-250MG
8-Benzyl-1,3,8-triaza-spiro[4.5]decan-4-one
8-Benzyl-1,3,8-triaza-spiro[4.5]decan-4-one
Formel: C₁₄H₁₉N₃O
Molecular Weight: 245,32 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13152416
CAS-Nummer: 170921-48-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR