Benzo[b]thiophen-4-ol
Lieferant: Apollo Scientific
|
Achtung
|
OR313024-1G
OR313024-250MG
APOSOR313024-1GEA
354
EUR
APOSOR313024-1G
APOSOR313024-250MG
Benzo[b]thiophen-4-ol
Benzo[b]thiophen-4-ol
Formel:
C₈H₆OS Molecular Weight: 150,198 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD01663547 CAS-Nummer: 3610-02-4 EINECS: 222-776-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...