N-Boc-4-(2-bromo-ethyl)-piperidine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 4-(2-bromoethyl)piperidine-1-carboxylate

OR313032-1G OR313032-5G
APOSOR313032-1GEA 114 EUR
APOSOR313032-1G APOSOR313032-5G
N-Boc-4-(2-bromo-ethyl)-piperidine 97%
N-Boc-4-(2-bromo-ethyl)-piperidine
Formel: C₁₂H₂₂BrNO₂
Molecular Weight: 292,213 g/mol
Schmelzpunkt: 39…42 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06410598
CAS-Nummer: 169457-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR