2-Methyl-1H-pyrrolo[2,3-c]pyridine 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR313057-1G
OR313057-250MG
APOSOR313057-1GEA
478
EUR
APOSOR313057-1G
APOSOR313057-250MG
2-Methyl-1H-pyrrolo[2,3-c]pyridine 97%
2-Methyl-1H-pyrrolo[2,3-c]pyridine
Formel:
C₈H₈N₂ Molecular Weight: 132,163 g/mol Schmelzpunkt: 183 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07776769 CAS-Nummer: 65645-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...