1,2,3,4-Tetrahydro-isoquinoline-5-carbonitrile 95%

Lieferant: Apollo Scientific
Achtung

OR313060-1G OR313060-250MG OR313060-100MG
APOSOR313060-1GEA 1450 EUR
APOSOR313060-1G APOSOR313060-250MG APOSOR313060-100MG
1,2,3,4-Tetrahydro-isoquinoline-5-carbonitrile 95%
1,2,3,4-Tetrahydro-isoquinoline-5-carbonitrile
Formel: C₁₀H₁₀N₂
Molecular Weight: 158,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07369985
CAS-Nummer: 215794-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR