(Pyrazin-2-yl)acetic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Pyrazin-2-yl)ethanoic acid, 2-(Carboxymethyl)pyrazine, 2-(Carboxymethyl)-1,4-diazine
OR313069-1G
OR313069-5G
APOSOR313069-1GEA
114
EUR
APOSOR313069-1G
APOSOR313069-5G
(Pyrazin-2-yl)acetic acid 97%
(Pyrazin-2-yl)acetic acid
Formel:
C₆H₆N₂O₂ Molecular Weight: 138,124 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD06796638 CAS-Nummer: 140914-89-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...