3-Chloro-5-methoxyaniline

Lieferant: Apollo Scientific
Achtung

OR313073-1G OR313073-5G OR313073-25G
APOSOR313073-1GEA 53.8 EUR
APOSOR313073-1G APOSOR313073-5G APOSOR313073-25G
3-Chloro-5-methoxyaniline
3-Chloro-5-methoxyaniline
Formel: C₇H₈ClNO
Molecular Weight: 157.598 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11035922
CAS-Nummer: 10272-06-7
EINECS: 233-614-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR