6-Hydroxy-5-methylpyridin-3-ylboronic acid 95%

Lieferant: Apollo Scientific
Achtung

OR315103-1G OR315103-250MG
APOSOR315103-1GEA 1060 EUR
APOSOR315103-1G APOSOR315103-250MG
6-Hydroxy-5-methylpyridin-3-ylboronic acid 95%
6-Hydroxy-5-methylpyridin-3-ylboronic acid
Formel: C₆H₈BNO₃
Molecular Weight: 152.944 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18909517
CAS-Nummer: 1356087-46-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR