6-Bromo-3-chlorobenzo[d]isoxazole

Lieferant: Apollo Scientific
Achtung

OR315304-1G OR315304-250MG
APOSOR315304-1GEA 504 EUR
APOSOR315304-1G APOSOR315304-250MG
6-Bromo-3-chlorobenzo[d]isoxazole
6-Bromo-3-chlorobenzo[d]isoxazole
Formel: C₇H₃BrClNO
Molecular Weight: 232,462 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12406141
CAS-Nummer: 1243389-57-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR